missing translation for 'onlineSavingsMsg'
Learn More

7-Azaindole, 98%

Catalog No. p-7044730
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0798303 1 g
AAL0798306 5 g
AAL0798314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0798303 Supplier Thermo Scientific Chemicals Supplier No. L0798303
Only null left

CAS: 271-63-6 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 271-63-6
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005606,MFCD00075874
InChI Key MVXVYAKCVDQRLW-UHFFFAOYSA-N
Synonym 7-azaindole, 1h-pyrrolo 2,3-b pyridine, 1,7-diazaindene, 7h-pyrrolo 2,3-b pyridine, 7-aza-1-pyrindine, 1,7-dideazapurine, 1 h-pyrrolo 2,3:b pyridine, 7-aza indole, unii-qx4465nr9t
PubChem CID 9222
IUPAC Name 1H-pyrrolo[2,3-b]pyridine
SMILES N1C=CC2=CC=CN=C12

Specifications

Melting Point 103°C to 107°C
Boiling Point 270°C
Quantity 1 g
Beilstein 109667
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 7-Azaindole
Hazard Category H315-H319
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-981-0
RTECSNumber UY8710000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only