missing translation for 'onlineSavingsMsg'
Learn More

7-Azaindole-5-carbonitrile, 97%

Catalog No. p-7030547
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH30237MD 250 mg
AAH3023703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH30237MD Supplier Thermo Scientific Chemicals Supplier No. H30237MD

CAS: 517918-95-5 | C8H5N3 | 143.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 517918-95-5
Molecular Formula C8H5N3
Molecular Weight (g/mol) 143.149
MDL Number MFCD06659684
InChI Key DRAQIXNADYAISI-UHFFFAOYSA-N
Synonym 1h-pyrrolo 2,3-b pyridine-5-carbonitrile, 5-cyano-7-azaindole, 7-azaindole-5-carbonitrile, 5-cyano-1h-pyrrolo 2,3-b pyridine, 5-canyo-1h-pyrrolo 2,3-b pyridine, pubchem16565, acmc-1avk2, ksc269e0f, 1h-pyrrolo 2,3-b pyridine-5-carbonitrile 9ci
PubChem CID 11788355
IUPAC Name 1H-pyrrolo[2,3-b]pyridine-5-carbonitrile
SMILES C1=CNC2=C1C=C(C=N2)C#N

Specifications

Melting Point 226°C to 230°C
Quantity 250 mg
Formula Weight 143.15
Percent Purity 97%
Chemical Name or Material 7-Azaindole-5-carbonitrile
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only