missing translation for 'onlineSavingsMsg'
Learn More

7-Azaindole-5-boronic acid pinacol ester, 97%

CAS: 754214-56-7 | C13H17BN2O2 | 244.10 g/mol

$85.62 - $243.62

Chemical Identifiers

CAS 754214-56-7
Molecular Formula C13H17BN2O2
Molecular Weight (g/mol) 244.10
MDL Number MFCD08060937
InChI Key UOXAMYZTYZLSCC-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 7-azaindole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, pyrrolo 2,3-b pyridine-5-boronic acid, pinacol ester, pyrrolo 2,3-b pyridin-5-ylboronic acid pinacol ester, zlchem 1246, pubchem16626, acmc-209szp, ksc641k4p
PubChem CID 24208789
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CN=C2NC=CC2=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH50044MD
View Documents
Thermo Scientific Chemicals
H50044MD
250 mg
Each for $85.62
Only null left
 
AAH5004403
View Documents
Thermo Scientific Chemicals
H5004403
1 g
Each for $243.62
Only null left
 
Description

Description

7-Azaindole-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Azaindole-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Soluble in methanol, ethanol and THF.

Notes
Air sensitive. Store away from oxidizing agents.
Specifications

Chemical Identifiers

754214-56-7
244.10
UOXAMYZTYZLSCC-UHFFFAOYSA-N
24208789
CC1(C)OB(OC1(C)C)C1=CN=C2NC=CC2=C1
C13H17BN2O2
MFCD08060937
5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 7-azaindole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, pyrrolo 2,3-b pyridine-5-boronic acid, pinacol ester, pyrrolo 2,3-b pyridin-5-ylboronic acid pinacol ester, zlchem 1246, pubchem16626, acmc-209szp, ksc641k4p
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine

Specifications

754214-56-7
C13H17BN2O2
250 mg
5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 7-azaindole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, pyrrolo 2,3-b pyridine-5-boronic acid, pinacol ester, pyrrolo 2,3-b pyridin-5-ylboronic acid pinacol ester, zlchem 1246, pubchem16626, acmc-209szp, ksc641k4p
UOXAMYZTYZLSCC-UHFFFAOYSA-N
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
24208789
97%
241°C to 245°C
MFCD08060937
Air Sensitive
Soluble in methanol,ethanol and THF.
CC1(C)OB(OC1(C)C)C1=CN=C2NC=CC2=C1
244.10
244.1
7-Azaindole-5-boronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only