missing translation for 'onlineSavingsMsg'
Learn More

7-Azaindole-3-carboxylic acid, 95%

Catalog No. aah5986603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5986603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5986603 Supplier Thermo Scientific Chemicals Supplier No. H5986603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 156270-06-3 | C8H6N2O2 | 162.148 g/mol

7-Azaindole-3-carboxylic acid is a reactant for synthesis of azaindol derivatives as new acrosin inhibitors and for preparation of triazoles via regioselective heterocyclizaiton reactions. It is also a reactant for synthesis of azaindolylcarboxy-endo-tropanamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Azaindole-3-carboxylic acid is a reactant for synthesis of azaindol derivatives as new acrosin inhibitors and for preparation of triazoles via regioselective heterocyclizaiton reactions. It is also a reactant for synthesis of azai

Chemical Identifiers

CAS 156270-06-3
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD07778360
InChI Key KYBIRFFGAIFLPM-UHFFFAOYSA-N
Synonym 1h-pyrrolo 2,3-b pyridine-3-carboxylic acid, 7-azaindole-3-carboxylic acid, 1h-pyrrolo 2,3-b pyridine-3-carboxylicacid, pubchem20644, acmc-209zs1, ksc173s3p, 7-azaindole-3-carboxylic ..., methyl 7-azaindole-3-carboxylic acid
PubChem CID 10154191
IUPAC Name 1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid
SMILES C1=CC2=C(NC=C2C(=O)O)N=C1

Specifications

Melting Point 205°C to 209°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 162.15
Percent Purity 95%
Chemical Name or Material 7-Azaindole-3-carboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only