missing translation for 'onlineSavingsMsg'
Learn More

7-Aminoheptanoic Acid 98.0+%, TCI America™

Catalog No. A03115G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03115G 5 g
A03111G 1 g
A031125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A03115G Supplier TCI America Supplier No. A03115G

Chemical Identifiers

CAS 929-17-9
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.202
MDL Number MFCD00008242
InChI Key XDOLZJYETYVRKV-UHFFFAOYSA-N
Synonym 7-aminoenanthic acid, heptanoic acid, 7-amino, omega-aminoenantic acid, aminoenanthic acid, amino-enanthylic acid, 7-amino-heptanoic acid, 7-aminohepentanoic acid, omega-aminoheptanoic acid, unii-b10hjx2t48, kyselina 7-aminoenanthova czech
PubChem CID 13580
IUPAC Name 7-aminoheptanoic acid
SMILES C(CCCN)CCC(=O)O

Specifications

Melting Point 189°C
Color White
Quantity 5 g
Formula Weight 145.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Aminoheptanoic Acid
RTECSNumber MJ1770000
TSCA No