missing translation for 'onlineSavingsMsg'
Learn More

7-Aminoheptanoic acid, 98%

Catalog No. AAH2751903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2751903 1 g
AAH2751906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2751903 Supplier Thermo Scientific Chemicals Supplier No. H2751903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 929-17-9 | C7H15NO2 | 145.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 929-17-9
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.202
MDL Number MFCD00008242
InChI Key XDOLZJYETYVRKV-UHFFFAOYSA-N
Synonym 7-aminoenanthic acid, heptanoic acid, 7-amino, omega-aminoenantic acid, aminoenanthic acid, amino-enanthylic acid, 7-amino-heptanoic acid, 7-aminohepentanoic acid, omega-aminoheptanoic acid, unii-b10hjx2t48, kyselina 7-aminoenanthova czech
PubChem CID 13580
IUPAC Name 7-aminoheptanoic acid
SMILES C(CCCN)CCC(=O)O

Specifications

Melting Point 194°C to 196°C
Quantity 1 g
Beilstein 906887
Formula Weight 145.19
Percent Purity 98%
Chemical Name or Material 7-Aminoheptanoic acid
EINECSNumber 213-197-5
RTECSNumber MJ1770000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only