missing translation for 'onlineSavingsMsg'
Learn More

7-Amino-4-(trifluoromethyl)coumarin, 97%

Catalog No. aaa1840003
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1840003 1 g
AAA1840001 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1840003 Supplier Thermo Scientific Chemicals Supplier No. A1840003
Only null left

CAS: 53518-15-3 | C10H6F3NO2 | 229.158 g/mol

7-Amino-4-(trifluoromethyl)coumarin is used in the synthesis of substrates for the fluorometric assay of proteolytic enzymes in biological fluids. It is suitable as laser dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Amino-4-(trifluoromethyl)coumarin is used in the synthesis of substrates for the fluorometric assay of proteolytic enzymes in biological fluids. It is suitable as laser dye.

Solubility
Soluble in dimethyl sulfoxide (25 mg/ml), dimethyl formamide (25 mg/ml), and methanol (25 mg/ml). Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 53518-15-3
Molecular Formula C10H6F3NO2
Molecular Weight (g/mol) 229.158
MDL Number MFCD00006858
InChI Key JBNOVHJXQSHGRL-UHFFFAOYSA-N
Synonym coumarin 151, 7-amino-4-trifluoromethyl coumarin, 7-amino-4-trifluoromethylcoumarin, 7-amino-4-trifluoromethyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-trifluoromethyl, 7-amino-4-trifluoromethyl chromen-2-one, 7-amino-4-trifluoromethyl-2-benzopyrone, 7-amino-4-trifluoromethyl-chromen-2-one, pubchem8665, maybridge1_007017
PubChem CID 100641
ChEBI CHEBI:51772
IUPAC Name 7-amino-4-(trifluoromethyl)chromen-2-one
SMILES C1=CC2=C(C=C1N)OC(=O)C=C2C(F)(F)F

Specifications

Melting Point 222°C to 223°C
Quantity 1 g
Beilstein 4456797
Solubility Information Soluble in dimethyl sulfoxide (25mg/ml),dimethyl formamide (25mg/ml),and methanol (25mg/ml). Insoluble in water.
Formula Weight 229.16
Percent Purity 99%
Chemical Name or Material 7-Amino-4-(trifluoromethyl)coumarin
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 258-599-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only