missing translation for 'onlineSavingsMsg'
Learn More

7-Amino-4-methylcoumarin 98.0+%, TCI America™

Catalog No. p-7132347
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07601G 1 g
M07605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M07601G Supplier TCI America Supplier No. M07601G
Only null left

Chemical Identifiers

CAS 26093-31-2
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00006868
InChI Key GLNDAGDHSLMOKX-UHFFFAOYSA-N
Synonym 7-amino-4-methylcoumarin, coumarin 120, 7-amino-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-methyl, coumarin, 7-amino-4-methyl, 4-methyl-7-aminocoumarin, 7-amino-4-methyl-chromen-2-one, unii-ocy3jct44x, ccris 4961, ocy3jct44x
PubChem CID 92249
ChEBI CHEBI:51771
IUPAC Name 7-amino-4-methyl-2H-chromen-2-one
SMILES CC1=CC(=O)OC2=CC(N)=CC=C12

Specifications

Melting Point 228°C
Quantity 1 g
Formula Weight 175.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Amino-4-methylcoumarin [for HPLC Labeling]