Learn More
Thermo Scientific Chemicals 7-Amino-4-methylcoumarin, 98%
CAS: 26093-31-2 | C10H9NO2 | 175.19 g/mol
$188.01 - $188.01
Chemical Identifiers
| CAS | 26093-31-2 |
|---|---|
| Molecular Formula | C10H9NO2 |
| Molecular Weight (g/mol) | 175.19 |
| MDL Number | MFCD00006868 |
| InChI Key | GLNDAGDHSLMOKX-UHFFFAOYSA-N |
| Synonym | 7-amino-4-methylcoumarin, coumarin 120, 7-amino-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-methyl, coumarin, 7-amino-4-methyl, 4-methyl-7-aminocoumarin, 7-amino-4-methyl-chromen-2-one, unii-ocy3jct44x, ccris 4961, ocy3jct44x |
| PubChem CID | 92249 |
| ChEBI | CHEBI:51771 |
| IUPAC Name | 7-amino-4-methylchromen-2-one |
| SMILES | CC1=CC(=O)OC2=CC(N)=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC218062500
|
Thermo Scientific Chemicals
218062500 |
250 mg | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 26093-31-2 | |
| 175.19 | |
| GLNDAGDHSLMOKX-UHFFFAOYSA-N | |
| 92249 | |
| 7-amino-4-methylchromen-2-one |
| C10H9NO2 | |
| MFCD00006868 | |
| 7-amino-4-methylcoumarin, coumarin 120, 7-amino-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-methyl, coumarin, 7-amino-4-methyl, 4-methyl-7-aminocoumarin, 7-amino-4-methyl-chromen-2-one, unii-ocy3jct44x, ccris 4961, ocy3jct44x | |
| CHEBI:51771 | |
| CC1=CC(=O)OC2=CC(N)=CC=C12 |
Specifications
| 26093-31-2 | |
| 100.0 | |
| Beige to Brown | |
| 97.5% min. (HPLC) | |
| C10H9NO2 | |
| 250 mg | |
| Solubility in water: slightly soluble. | |
| CC1=CC(=O)OC2=CC(N)=CC=C12 | |
| 175.19 | |
| CHEBI:51771 | |
| 98% | |
| Crystalline Powder |
| 97.5 | |
| 223.0°C to 226.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00006868 | |
| 7-amino-4-methylcoumarin, coumarin 120, 7-amino-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-methyl, coumarin, 7-amino-4-methyl, 4-methyl-7-aminocoumarin, 7-amino-4-methyl-chromen-2-one, unii-ocy3jct44x, ccris 4961, ocy3jct44x | |
| GLNDAGDHSLMOKX-UHFFFAOYSA-N | |
| 7-amino-4-methylchromen-2-one | |
| 92249 | |
| 175.19 | |
| Extra Pure | |
| 7-Amino-4-methylcoumarin |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 247-454-8
RUO – Research Use Only