missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Amino-2-benzoxazolinone 95.0+%, TCI America™
$74.56 - $205.05
Chemical Identifiers
| CAS | 81282-60-2 |
|---|---|
| Molecular Formula | C7H6N2O2 |
| Molecular Weight (g/mol) | 150.137 |
| MDL Number | MFCD09753616 |
| InChI Key | CLCPWTXGFUIRJE-UHFFFAOYSA-N |
| Synonym | 7-Amino-2-benzoxazolone, 7-Amino-2-hydroxybenzoxazole |
| PubChem CID | 11147814 |
| IUPAC Name | 7-amino-3H-1,3-benzoxazol-2-one |
| SMILES | C1=CC2=C(C(=C1)N)OC(=O)N2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A2806250MG
|
TCI America
A2806250MG |
250 mg |
Each for $74.56
|
|
|||||
|
A28061G
|
TCI America
A28061G |
1 g |
Each for $205.05
|
|
|||||
Chemical Identifiers
| 81282-60-2 | |
| 150.137 | |
| CLCPWTXGFUIRJE-UHFFFAOYSA-N | |
| 11147814 | |
| C1=CC2=C(C(=C1)N)OC(=O)N2 |
| C7H6N2O2 | |
| MFCD09753616 | |
| 7-Amino-2-benzoxazolone, 7-Amino-2-hydroxybenzoxazole | |
| 7-amino-3H-1,3-benzoxazol-2-one |
Specifications
| 81282-60-2 | |
| C7H6N2O2 | |
| 250 mg | |
| CLCPWTXGFUIRJE-UHFFFAOYSA-N | |
| 7-amino-3H-1,3-benzoxazol-2-one | |
| 11147814 | |
| ≥95.0% (GC,T) | |
| 7-Amino-2-benzoxazolinone |
| Yellow | |
| MFCD09753616 | |
| 7-Amino-2-benzoxazolone, 7-Amino-2-hydroxybenzoxazole | |
| C1=CC2=C(C(=C1)N)OC(=O)N2 | |
| 150.137 | |
| 150.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No