missing translation for 'onlineSavingsMsg'
Learn More

7,8-Dihydroxy-4-methylcoumarin 98.0+%, TCI America™

Catalog No. p-7128466
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47931G 1 g
D47935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47931G Supplier TCI America Supplier No. D47931G
Only null left

Chemical Identifiers

CAS 2107-77-9
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00016972
InChI Key NWQBYMPNIJXFNQ-UHFFFAOYSA-N
Synonym 7,8-dihydroxy-4-methylcoumarin, 4-methyldaphnetin, 7,8-dihydroxy-4-methyl-2h-chromen-2-one, 4-methyldaphnetine, 4-methyl-7,8-dihydroxycoumarin, 2h-1-benzopyran-2-one, 7,8-dihydroxy-4-methyl, coumarin, 7,8-dihydroxy-4-methyl, 7,8-dihydroxy-4-methyl-2h-1-benzopyran-2-one, 7,8-dihydroxy-4-methyl-chromen-2-one, 7,8-dihydroxy-4-methyl-2-benzopyrone
PubChem CID 5355836
IUPAC Name 7,8-dihydroxy-4-methyl-2H-chromen-2-one
SMILES CC1=CC(=O)OC2=C(O)C(O)=CC=C12

Specifications

Melting Point 243°C
Quantity 1 g
Formula Weight 192.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7,8-Dihydroxy-4-methylcoumarin
RTECSNumber GN6385000
TSCA No
Recommended Storage Refrigerator