missing translation for 'onlineSavingsMsg'
Learn More

7,8-Dihydroxy-4-methylcoumarin, 97%

Catalog No. AAL14069MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14069MC 100 mg
AAL14069ME 500 mg
AAL1406903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL14069MC Supplier Thermo Scientific Chemicals Supplier No. L14069MC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2107-77-9 | C10H8O4 | 192.17 g/mol

Can be used as a fluorescent sensor to monitor the consumption of a boronic acid in a Suzuki cross-coupling reaction. Use a long wave UV lamp.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2107-77-9
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00016972
InChI Key NWQBYMPNIJXFNQ-UHFFFAOYSA-N
Synonym 7,8-dihydroxy-4-methylcoumarin, 4-methyldaphnetin, 7,8-dihydroxy-4-methyl-2h-chromen-2-one, 4-methyldaphnetine, 4-methyl-7,8-dihydroxycoumarin, 2h-1-benzopyran-2-one, 7,8-dihydroxy-4-methyl, coumarin, 7,8-dihydroxy-4-methyl, 7,8-dihydroxy-4-methyl-2h-1-benzopyran-2-one, 7,8-dihydroxy-4-methyl-chromen-2-one, 7,8-dihydroxy-4-methyl-2-benzopyrone
PubChem CID 5355836
SMILES CC1=CC(=O)OC2=C(O)C(O)=CC=C12

Specifications

Melting Point 241°C to 244°C
Quantity 100 mg
Beilstein 177613
Sensitivity Light sensitive
IUPAC Name 7,8-dihydroxy-4-methyl-2H-chromen-2-one
Formula Weight 192.17
Percent Purity 97%
Chemical Name or Material 7,8-Dihydroxy-4-methylcoumarin
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-290-4
RTECSNumber GN6385000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only