missing translation for 'onlineSavingsMsg'
Learn More

7,8-Benzoquinoline, 97%, Thermo Scientific™

Catalog No. AC154070010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154070010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC154070010 Supplier Thermo Scientific Chemicals Supplier No. 154070010

Chemical Identifiers

CAS 230-27-3
Molecular Formula C13H9N
Molecular Weight (g/mol) 179.22
MDL Number MFCD00004984
InChI Key WZJYKHNJTSNBHV-UHFFFAOYSA-N
Synonym benzo h quinoline, 7,8-benzoquinoline, 4-azaphenanthrene, 1-naphthoquinoline, benzoquinoline, alpha-naphthoquinoline, alpha-benzoquinoline, 7,8-bnzoquinoline, .alpha.-benzoquinoline
PubChem CID 9191
IUPAC Name benzo[h]quinoline
SMILES C1=CC=C2C(=C1)C=CC3=C2N=CC=C3

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 48°C to 52°C
Color White to Brown
Boiling Point 338°C (719.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 20, 463
Formula Weight 179.22
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 7, 8-Benzoquinoline, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-937-

RUO – Research Use Only