missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7,7-Dichlorobicyclo[4.1.0]heptane 98.0+%, TCI America™
$76.43 - $372.81
Chemical Identifiers
| CAS | 823-69-8 |
|---|---|
| Molecular Formula | C7H10Cl2 |
| Molecular Weight (g/mol) | 165.057 |
| MDL Number | MFCD00466186 |
| InChI Key | MBPSCGRETWPLSI-UHFFFAOYSA-N |
| PubChem CID | 244375 |
| IUPAC Name | 7,7-dichlorobicyclo[4.1.0]heptane |
| SMILES | C1CCC2C(C1)C2(Cl)Cl |
Chemical Identifiers
| 823-69-8 | |
| 165.057 | |
| MBPSCGRETWPLSI-UHFFFAOYSA-N | |
| 7,7-dichlorobicyclo[4.1.0]heptane |
| C7H10Cl2 | |
| MFCD00466186 | |
| 244375 | |
| C1CCC2C(C1)C2(Cl)Cl |
Specifications
| 823-69-8 | |
| 80°C | |
| MFCD00466186 | |
| MBPSCGRETWPLSI-UHFFFAOYSA-N | |
| 7,7-dichlorobicyclo[4.1.0]heptane | |
| 244375 | |
| ≥98.0% (GC) | |
| 7,7-Dichlorobicyclo[4.1.0]heptane |
| Yellow | |
| C7H10Cl2 | |
| 5 g | |
| C1CCC2C(C1)C2(Cl)Cl | |
| 165.057 | |
| 165.06 | |
| Liquid |
Safety and Handling
TSCA : No