missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7,7,8,8-Tetracyanoquinodimethane (purified by sublimation) 99.0+%, TCI America™
$126.76 - $380.27
Chemical Identifiers
| CAS | 1518-16-7 |
|---|---|
| Molecular Formula | C12H4N4 |
| Molecular Weight (g/mol) | 204.192 |
| MDL Number | MFCD00011664 |
| InChI Key | PCCVSPMFGIFTHU-UHFFFAOYSA-N |
| Synonym | 7,7,8,8-tetracyanoquinodimethane, tcnq, tetracyanoquinodimethane, tetracyanoquinodimethan, 2,2'-cyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tetracyano-p-quinodimethane, tetracyanoquinodimethan e, quinodimethan, tetracyano, propanedinitrile, 2,2'-2,5-cyclohexadiene-1,4-diylidene bis, 7,7,8,8-tetracyano-p-quinodimethane |
| PubChem CID | 73697 |
| ChEBI | CHEBI:52445 |
| IUPAC Name | 2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile |
| SMILES | C1=CC(=C(C#N)C#N)C=CC1=C(C#N)C#N |
Chemical Identifiers
| 1518-16-7 | |
| 204.192 | |
| PCCVSPMFGIFTHU-UHFFFAOYSA-N | |
| 73697 | |
| 2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile |
| C12H4N4 | |
| MFCD00011664 | |
| 7,7,8,8-tetracyanoquinodimethane, tcnq, tetracyanoquinodimethane, tetracyanoquinodimethan, 2,2'-cyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tetracyano-p-quinodimethane, tetracyanoquinodimethan e, quinodimethan, tetracyano, propanedinitrile, 2,2'-2,5-cyclohexadiene-1,4-diylidene bis, 7,7,8,8-tetracyano-p-quinodimethane | |
| CHEBI:52445 | |
| C1=CC(=C(C#N)C#N)C=CC1=C(C#N)C#N |
Specifications
| 1518-16-7 | |
| C12H4N4 | |
| 1 g | |
| 7,7,8,8-tetracyanoquinodimethane, tcnq, tetracyanoquinodimethane, tetracyanoquinodimethan, 2,2'-cyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tetracyano-p-quinodimethane, tetracyanoquinodimethan e, quinodimethan, tetracyano, propanedinitrile, 2,2'-2,5-cyclohexadiene-1,4-diylidene bis, 7,7,8,8-tetracyano-p-quinodimethane | |
| C1=CC(=C(C#N)C#N)C=CC1=C(C#N)C#N | |
| 204.192 | |
| CHEBI:52445 | |
| ≥99.0% (HPLC,N) | |
| 7,7,8,8-Tetracyanoquinodimethane (purified by sublimation) |
| 289°C | |
| MFCD00011664 | |
| 3439 | |
| PCCVSPMFGIFTHU-UHFFFAOYSA-N | |
| 2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile | |
| 73697 | |
| 204.19 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1798
RTECSNumber : GU4850000
TSCA : Yes