missing translation for 'onlineSavingsMsg'
Learn More

7,7,8,8-Tetracyanoquinodimethane (purified by sublimation) 99.0+%, TCI America™

Catalog No. T30341G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30341G 1 g
T30345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30341G Supplier TCI America Supplier No. T30341G

Chemical Identifiers

CAS 1518-16-7
Molecular Formula C12H4N4
Molecular Weight (g/mol) 204.192
MDL Number MFCD00011664
InChI Key PCCVSPMFGIFTHU-UHFFFAOYSA-N
Synonym 7,7,8,8-tetracyanoquinodimethane, tcnq, tetracyanoquinodimethane, tetracyanoquinodimethan, 2,2'-cyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tetracyano-p-quinodimethane, tetracyanoquinodimethan e, quinodimethan, tetracyano, propanedinitrile, 2,2'-2,5-cyclohexadiene-1,4-diylidene bis, 7,7,8,8-tetracyano-p-quinodimethane
PubChem CID 73697
ChEBI CHEBI:52445
IUPAC Name 2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile
SMILES C1=CC(=C(C#N)C#N)C=CC1=C(C#N)C#N

Specifications

Melting Point 289°C
Quantity 1 g
UN Number 3439
Formula Weight 204.19
Percent Purity ≥99.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 7,7,8,8-Tetracyanoquinodimethane (purified by sublimation)
EINECSNumber (3)-1798
RTECSNumber GU4850000
TSCA Yes