missing translation for 'onlineSavingsMsg'
Learn More

7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydrobenzo[b]furan, 95%, Thermo Scientific™

Catalog No. p-3882121
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC00939CB 250mg Amber Glass Bottle
CC00939DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC00939CB Supplier Thermo Scientific Chemicals Supplier No. CC00939CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 934586-50-2
Molecular Formula C14H19BO3
Molecular Weight (g/mol) 246.113
MDL Number MFCD11109314
InChI Key HCAAUTYLSSDHFV-UHFFFAOYSA-N
Synonym 2-2,3-dihydrobenzofuran-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydrobenzo b furan, 2,3-dihydrobenzofuran-7-boronic acid, pinacol ester, 2-2,3-dihydro-1-benzofuran-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2,3-dihydro-benzofuran, 7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1-benzofuran, 2,3-dihydrobenzo b furan-7-boronic acid pinacol ester, 2-2,3-dihydrobenzo 2,3-b furan-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 43811027
IUPAC Name 2-(2,3-dihydro-1-benzofuran-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C3C(=CC=C2)CCO3

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 246.11
Percent Purity 95%
Chemical Name or Material 7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydrobenzo[b]furan
Hazard Category Warning