missing translation for 'onlineSavingsMsg'
Learn More

7-(2-Hydroxypropyl)theophylline 98.0+%, TCI America™

Catalog No. H14305G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14305G 5 g
H143025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14305G Supplier TCI America Supplier No. H14305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 603-00-9
Molecular Formula C10H14N4O3
Molecular Weight (g/mol) 238.25
MDL Number MFCD00022833
InChI Key KYHQZNGJUGFTGR-UHFFFAOYNA-N
Synonym 7-(2-Hydroxypropyl)-1,3-dimethylxanthine, Proxyphylline
PubChem CID 4977
IUPAC Name 7-(2-hydroxypropyl)-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
SMILES CC(O)CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 5 g
Formula Weight 238.25
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 7-(2-Hydroxypropyl)theophylline
RTECSNumber XH5907500
TSCA No