Learn More
CDS Empore™ 6602 E4tip™ GB+C18 Membrane, 3-Layer, 200 μL
Catalog No. 13110088
Change view
Click to view available options
Quantity:
25/Pk.
96/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13110088 96/Cs.
13110087 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13110088 Supplier CDS Empore™ Supplier No. 70201930035
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
E3technology™ is an efficient, effective and economical approach for proteomics cell lysis, providing digestion ready samples in mere minutes, completely eliminating various detergents and interferences.
  • Efficient: < 15 min. hands-on time
  • Effective: compatible with a variety of upstream cell lysis conditions (e.g., urea, SDS, RIPA, TFA, etc.)
  • Economical
  • Robust: zero technical barrier to even entry-level biomedical scientists
  • Versatile: E3tip, E3filter, E3cartridge, and E3plate to satisfy different sample volumes, concentration, quantities, and the need for automation
  • Stress-free: no concerns of solution pH, protein concentration, protein-bead ratios, beads stick to tube walls and surfaces, cross-contaminations, etc.
  • E4technology: enhanced 'single vessel' approach for low-cell proteomics
  • Loading capacity: up to 50,000 cells
Specifications
Bed Weight 50 mg
For Use With (Equipment) Vacuum Manifolds or Automated SPE Equipment
Particle Size 30 μm
Type SPE Tips
Volume (Metric) 200 μL
Quantity 96/Cs.