Learn More
CDS Empore™ 6602 E4tip™ GB+C18 Membrane, 3-Layer, 10 μL
E3technology™ is an efficient, effective and economical approach for proteomics cell lysis, providing digestion ready samples in mere minutes, completely eliminating various detergents and interferences.
Supplier:  CDS Empore™ 70201930047EA
 View more versions of this product
Catalog No. 13110085
Only null left
Description
Description
  • Efficient: < 15 min. hands-on time
  • Effective: compatible with a variety of upstream cell lysis conditions (e.g., urea, SDS, RIPA, TFA, etc.)
  • Economical
  • Robust: zero technical barrier to even entry-level biomedical scientists
  • Versatile: E3tip, E3filter, E3cartridge, and E3plate to satisfy different sample volumes, concentration, quantities, and the need for automation
  • Stress-free: no concerns of solution pH, protein concentration, protein-bead ratios, beads stick to tube walls and surfaces, cross-contaminations, etc.
  • E4technology: enhanced 'single vessel' approach for low-cell proteomics
  • Loading capacity: up to 10,000 cells
Specifications
Chemical Identifiers
Vacuum Manifolds or Automated SPE Equipment
30 μm
SPE Tips
10 μL
Specifications
Vacuum Manifolds or Automated SPE Equipment
SPE Tips
25/Pk.
30 μm
10 μL
Product Suggestions
Product Suggestions