Learn More
Celltreat Scientific Products 60mm x 15mm Petri Dish w/Grip Ring, Sterile
Supplier: Celltreat Scientific Products 229663
CELLTREAT Scientific Products 60mm x 15mm Non-Treated Petri Dish. Dishes have thicker and more durable plastic than other Petri Dishes. Vented lids contain molded spacers to allow gas exchange. Working volume of 7-8mL and growth area of 21cm. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 10/re-sealable bag, 500/case.