Learn More
YSI™ 60520 Pro Series DO Cable
Catalog No. 15177472
Change view
Click to view available options
Length (English) Cable:
3.2 ft.
13 ft.
32 ft.
65 ft.
328 ft.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Cable
15177472 65 ft.
15177470 13 ft.
15176217 328 ft.
15177471 32 ft.
1339889 3.2 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 15177472 Supplier YSI™ Supplier No. 6052020
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Compatible with the YSI Pro Plus or Pro20 handhelds
Easy user-replaceable sensor allows for quick sensor replacements, if needed. No tools necessary. One-year warranty.Measure pH quickly and easily with the 1001 sensor.The 605101 pH Sensor can be used with the YSI Professional Plus Multiparameter Handheld Instrument. Requires Cable/Probe Assembly.Item is heat packed in the winter months. Shipment must be made via 2-day or next day carrier during that time information.
Cables over 1-meter also come with the 603062 cable management kit.
Order Info
Cables do NOT come with the sensor
Specifications
Type Cable Assembly
For Use With (Application) Measure temperature and DO
Length (English) Cable 65 ft.
Length (Metric) Cable 20 m
Warranty 2 Years