missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ ASSY, Sample Collector .50LD Z
Description
This is a spare part for the 310 Genetic Analyzer.
Specifications
Specifications
| Quantity | 1 unit |
| For Use With (Equipment) | 310 Genetic Analyzer |
| Product Type | Sample Collector Assembly |
| Shipping Condition | Room Temperature |
| Content And Storage | Store at room temperature. |
For Research Use Only. Not for use in diagnostic procedures.