Learn More
Chemglass Inc 600mL Freeze Dry Flask, Crosses to Labconco P/N 7542800
Catalog No. CG161106
Encompass
Flat bottom borosilicate flask can be used with Labconco and Virtis freeze dryers. Wide mouth design provides easy access sample. PLEASE NOTE: FLASKS ONLY. NOT SUPPLIED WITH RUBBER TOP, FILTER OR O-RING.Note: Labconco and Virtis are not related to or affiliated with Chemglass, and these manufacturers have not approved or validated these products for use on their equipment.
Catalog No. CG161106 Supplier Chemglass Inc Supplier No. CG161106
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Flat bottom borosilicate flask can be used with Labconco and Virtis freeze dryers. Wide mouth design provides easy access sample. PLEASE NOTE: FLASKS ONLY. NOT SUPPLIED WITH RUBBER TOP, FILTER OR O-RING.Note: Labconco and Virtis are not related to or affiliated with Chemglass, and these manufacturers have not approved or validated these products for use on their equipment.