missing translation for 'onlineSavingsMsg'
Learn More

3M™ 6000 Series Half Face Mask Respirator

Catalog No. p-56499
Change view
Click to view available options
Size:
Small
Medium
Large
For Use With (Application):
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
Product Type:
Half-Mask Respirator
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size For Use With (Application) Product Type
179869A Small Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding Half-Mask Respirator
179869B Medium Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding Half-Mask Respirator
179869C Large Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding Half-Mask Respirator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 179869A Supplier 3M™ Supplier No. 7000001932
Only null left

Requires

Facepiece requires compatible cartridges and/or filters. (Sold separately.)

Half-face mask respirators 6000 Series are economical, low-maintenance, and extremely lightweight. Probed reusable respirator assembly 6000Q series are used for quantitative fit purposes only.

Combines features of both reusable and disposable respirators to provide effective protection. 3M™ Half Face Mask Respirator 6000 Series are economical, low-maintenance, simple-to-handle, and extremely lightweight.

  • Reusable half masks are economical, low-maintenance, simple-to-handle, and extremely lightweight
  • Used with 3M™ Supplied Air Systems
  • Bayonet Connection System allows connection to a broad range of twin lightweight filters to protect against gases, vapors, and particulates
  • Twin filter design provides lower breathing resistance
  • Probed reusable respirator assembly 6000 Series used for quantitative fit testing (17-987-3A & -3B)

Order Info

Probed models for fit test only—not for use as a respirator. Cartridge/filter combinations sold separately.

Specifications

Certifications (EN) EN 140:1998
Certifications (NIOSH) NIOSH 42 CFR Part 84
Connection Type Bayonet
Color Gray
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Small
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
For Use With (Equipment) Cartridge 6000 Series, Filter 2000, 2200, 7000 or 5000 Series, or Dual Airline System
Includes Reclosable storage bag
Model 6100
Suspension Type 4-point
Show More Show Less