missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ 60 Gallon Red Biohazard Waste Bag

Catalog No. 17988503
Change view
Click to view available options
Capacity (English):
60 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17988503 60 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17988503 Supplier Eagle™ Supplier No. 1693

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biohazard waste bag for holding or consolidating larger amounts of biological waste or contaminated materials

  • Designed to fit Eagle™ drums
  • Once bag is full and ready for disposal, seal bag in place and close drum using its lid and lever-lock
  • Red color bag is easily identifiable as biohazard bags
  • Drums can be black, yellow, blue or red
  • Printing includes English, Spanish and French translation along with international recognizable biohazard symbol
  • Material: linear low density polyethylene (LLDPE)
  • Thickness: 1.2 mil
  • High level of flexibility and shear resistance can be attained without leaks
  • Meets ASTM D 1709-01 and ASTM D 1922-00a specifications
  • Tubular construction, single side seal and star seal bottom provide stable, leak proof bag
  • 100/Pk.

Specifications

Capacity (English) 60 gal.
Color Red
Quantity 100/Pk.
Autoclavable Not Autoclavable
Liner Material LDPE
For Use With (Equipment) Eagle™ Drums
Width (English) 38 in.
Length (English) 58 in.
Thickness 1.2 mil
Height (English) 38 in.
Material Polyethylene
Product Type Biohazard Waste Bags
Show More Show Less