Learn More
Celltreat Scientific Products 6 Well Glass Bottom Tissue Culture Plate, 20mm Glass, Sterile
Supplier: Celltreat Scientific Products 229107
CELLTREAT Scientific Products 6 Well Glass Bottom Plate with Lid. Glass bottom plates have the same great features as standard tissue culture plates with the added benefit of borosilicate glass bottom microwells. Convenient live cell viewing without needing to transfer. Optically clear borosilicate glass for exceptional viewing. Applications include: cell culture and imaging, live cell observation and specialty microscopy such as confocal high resolution and contrast. Traditional footprints cause minimal disruption in cell culture techniques. Constructed with noncytotoxic medical adhesive. Round cover glass also available for use with glass bottom plates. 6 Well Tissue Culture Plate has 20mm glass bottom. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Individually packed sterile, 5/case.