Learn More
Alkali Scientific ASI™ 6-Well Confocal Plate (35m), Clear, Adhesion Type, Sterile, 4/Cs, PS/Flux, Treated (Positive-Charged/ Surface Coated), 50/Cs, 22 x 22 x 0.13mm, Hole 3.14mm
Catalog No. 503028884
Encompass
ASI™confocal products allow researchers to acquire high resolution microscopic images of cells in 35 mm culture dishes or 6 well plates. Low auto-fluorescence glass and FLux were used, ideal for uses of these products are confocal microscopy, phase contrast microscopy, live cell imaging and micro-manipulations. Surface materials for confocal products:Glass: naturally charged, good growth surface for adherent cellsFLux: surface modified plastic film (188 m thick), optimal for confocal microscopy Bottom materials: Glass/FLux (no surface treatment for glass bottom) Dish color: Clear Dish size: 35 Plate size: 6well Plate Hole size: 13 / 20 / 25 Non pyrogenic Non cytotoxic DNase / RNase free Human DNA free
Catalog No. 503028884 Supplier Alkali Scientific Supplier No. TCP24
ASI™confocal products allow researchers to acquire high resolution microscopic images of cells in 35 mm culture dishes or 6 well plates. Low auto-fluorescence glass and FLux were used, ideal for uses of these products are confocal microscopy, phase contrast microscopy, live cell imaging and micro-manipulations. Surface materials for confocal products:Glass: naturally charged, good growth surface for adherent cellsFLux: surface modified plastic film (188 m thick), optimal for confocal microscopy Bottom materials: Glass/FLux (no surface treatment for glass bottom) Dish color: Clear Dish size: 35 Plate size: 6well Plate Hole size: 13 / 20 / 25 Non pyrogenic Non cytotoxic DNase / RNase free Human DNA free