missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrandTech™ BRAND™ BIO-CERT™ CELL CULTURE QUALITY 2-in-1 Culture Inserts for 6-well Plates, 8 μL Pore Size, PET Membrane
Sterile inserts for 6-well plates, featuring cellGrade™ plus surface treatment for uniform cell growth. Designed with feet for standing or a removable hook for hanging, offering versatile use in cell culture applications.
Supplier: BrandTech™ BRAND™ 782757
Description
- Suitable for 2D and 3D cell cultures
- TC treated with cellGrade™ plus surface treatment for enhanced cell growth
- Highly transparent PET membrane ideal for imaging
- Sterile according to ISO 11137, SAL 10-6
- Produced in a Class 8 cleanroom per ISO 14644-1
- Non-cytotoxic, meeting ISO 10993-5 standards
- BIO-CERT™ CELL CULTURE QUALITY means inserts are free of pyrogens, endotoxins, human DNA, RNase, and DNase
- Diameter size of 24.8 mm for an ideal fit for 6-well plates
Specifications
| No Barcode | |
| Sterile according to ISO 11137, SAL 10-6, Non-cytotoxic according to ISO 10993-5, pyrogen and endotoxin free to 0.01EU/mL | |
| Without Lid | |
| 4.83 cm2 | |
| Not Autoclavable | |
| 8 μm | |
| Clear | |
| Microplate Insert | |
| Polystyrene | |
| 6 | |
| Sterile | |
| Round, Flat Bottom (F-well) | |
| 2D and 3D cell culture, co-culture, migration, chemotaxis, TEER assays, and air-lift culture | |
| Yes | |
| 11 mm |
| No | |
| Yes | |
| Ambient | |
| PET | |
| Multi-Pack Blister, 24/Pk. | |
| cellGrade plus | |
| Single-use | |
| 6-well plate | |
| 3000 μL | |
| 800 to 3000 μL | |
| cellGrade Plus | |
| Yes | |
| 24 | |
| Clear | |
| 24.8 mm |