missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(Trifluoromethyl)uracil, 97%, Thermo Scientific™
$97.45 - $312.47
Chemical Identifiers
| CAS | 672-45-7 |
|---|---|
| Molecular Formula | C5H3F3N2O2 |
| Molecular Weight (g/mol) | 180.09 |
| MDL Number | MFCD01011762 |
| InChI Key | IROWWTVZNHKLLE-UHFFFAOYSA-N |
| Synonym | 6-trifluoromethyl uracil, 2,4-dihydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidine-2,4-diol, uracil, 6-trifluoromethyl, 6-trifluoromethyl-pyrimidine-2,4-diol, 6-trifluoromethyl pyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 6-trifluoromethyl, 6-trifluoromethyluracil, 6-trifluoromethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 6-trifluoromethyl-1h-pyrimidine-2,4-dione |
| PubChem CID | 233890 |
| SMILES | FC(F)(F)C1=CC(=O)NC(=O)N1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 672-45-7 | |
| 180.09 | |
| IROWWTVZNHKLLE-UHFFFAOYSA-N | |
| 233890 |
| C5H3F3N2O2 | |
| MFCD01011762 | |
| 6-trifluoromethyl uracil, 2,4-dihydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidine-2,4-diol, uracil, 6-trifluoromethyl, 6-trifluoromethyl-pyrimidine-2,4-diol, 6-trifluoromethyl pyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 6-trifluoromethyl, 6-trifluoromethyluracil, 6-trifluoromethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 6-trifluoromethyl-1h-pyrimidine-2,4-dione | |
| FC(F)(F)C1=CC(=O)NC(=O)N1 |
Specifications
| 672-45-7 | |
| MFCD01011762 | |
| 6-trifluoromethyl uracil, 2,4-dihydroxy-6-trifluoromethylpyrimidine, 6-trifluoromethyl pyrimidine-2,4-diol, uracil, 6-trifluoromethyl, 6-trifluoromethyl-pyrimidine-2,4-diol, 6-trifluoromethyl pyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 6-trifluoromethyl, 6-trifluoromethyluracil, 6-trifluoromethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 6-trifluoromethyl-1h-pyrimidine-2,4-dione | |
| FC(F)(F)C1=CC(=O)NC(=O)N1 | |
| 180.09 | |
| 180.08 | |
| Powder |
| C5H3F3N2O2 | |
| 1 g | |
| IROWWTVZNHKLLE-UHFFFAOYSA-N | |
| 6-(trifluoromethyl)-1H-pyrimidine-2,4-dione | |
| 233890 | |
| 97% | |
| 6- (Trifluoromethyl)uracil |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures