missing translation for 'onlineSavingsMsg'
Learn More

6-(Trifluoromethyl)pyridine-2-carboxylic acid, 98%

CAS: 131747-42-7 | C7H4F3NO2 | 191.11 g/mol

Supplier:  Thermo Scientific Chemicals H6313803

 View more versions of this product

Catalog No. AAH6313803


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

6-(Trifluoromethyl)pyridine-2-carboxylic acid
154°C to 158°C
C7H4F3NO2
1 g
6-trifluoromethyl picolinic acid, 6-trifluoromethyl pyridine-2-carboxylic acid, 2-trifluoromethyl-6-pyridinecarboxylic acid, 6-trifluoromethyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 6-trifluoromethyl, 6-trifluoromethyl pyridine-2-carboxylicacid, 6-trifluoromethyl-pyridine-2-carboxylic acid, pubchem15170, 2-trifluoromethyl-6-pyridinecarboxylicacid, acmc-1ca7g
OC(=O)C1=NC(=CC=C1)C(F)(F)F
191.11
191.11
Solid
131747-42-7
White
MFCD02180819
UN2811
OKBHXGBLXDNJJD-UHFFFAOYSA-N
6-(trifluoromethyl)pyridine-2-carboxylic acid
14761453
98%

Chemical Identifiers

131747-42-7
191.11
OKBHXGBLXDNJJD-UHFFFAOYSA-N
14761453
OC(=O)C1=NC(=CC=C1)C(F)(F)F
C7H4F3NO2
MFCD02180819
6-trifluoromethyl picolinic acid, 6-trifluoromethyl pyridine-2-carboxylic acid, 2-trifluoromethyl-6-pyridinecarboxylic acid, 6-trifluoromethyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 6-trifluoromethyl, 6-trifluoromethyl pyridine-2-carboxylicacid, 6-trifluoromethyl-pyridine-2-carboxylic acid, pubchem15170, 2-trifluoromethyl-6-pyridinecarboxylicacid, acmc-1ca7g
6-(trifluoromethyl)pyridine-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only