Learn More
6-(Trifluoromethyl)nicotinic acid, 97%
CAS: 231291-22-8 | C7H3F3NO2 | 190.10 g/mol
$128.34 - $185.36
Chemical Identifiers
| CAS | 231291-22-8 |
|---|---|
| Molecular Formula | C7H3F3NO2 |
| Molecular Weight (g/mol) | 190.10 |
| MDL Number | MFCD00792430 |
| InChI Key | JNYLMODTPLSLIF-UHFFFAOYSA-M |
| Synonym | 6-trifluoromethyl nicotinic acid, 6-trifluoromethyl pyridine-3-carboxylic acid, 6-trifluoromethylnicotinic acid, 6-trifluoromethyl-nicotinic acid, 2-trifluoromethyl-5-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 6-trifluoromethyl, 2-trifluoromethyl pyridine-5-carboxylic acid, 6-trifluoromethyl-3-pyridinecarboxylic acid |
| PubChem CID | 2777551 |
| IUPAC Name | 6-(trifluoromethyl)pyridine-3-carboxylic acid |
| SMILES | [O-]C(=O)C1=CC=C(N=C1)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL20054MD
|
Thermo Scientific Chemicals
L20054MD |
250 mg |
Each for $128.34
|
|
|||||
|
AAL2005403
|
Thermo Scientific Chemicals
L2005403 |
1 g |
Each for $185.36
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 231291-22-8 | |
| 190.10 | |
| JNYLMODTPLSLIF-UHFFFAOYSA-M | |
| 2777551 | |
| [O-]C(=O)C1=CC=C(N=C1)C(F)(F)F |
| C7H3F3NO2 | |
| MFCD00792430 | |
| 6-trifluoromethyl nicotinic acid, 6-trifluoromethyl pyridine-3-carboxylic acid, 6-trifluoromethylnicotinic acid, 6-trifluoromethyl-nicotinic acid, 2-trifluoromethyl-5-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 6-trifluoromethyl, 2-trifluoromethyl pyridine-5-carboxylic acid, 6-trifluoromethyl-3-pyridinecarboxylic acid | |
| 6-(trifluoromethyl)pyridine-3-carboxylic acid |
Specifications
| 231291-22-8 | |
| C7H3F3NO2 | |
| 250 mg | |
| JNYLMODTPLSLIF-UHFFFAOYSA-M | |
| 6-(trifluoromethyl)pyridine-3-carboxylic acid | |
| 2777551 | |
| 97% |
| 191°C to 192°C | |
| MFCD00792430 | |
| 6-trifluoromethyl nicotinic acid, 6-trifluoromethyl pyridine-3-carboxylic acid, 6-trifluoromethylnicotinic acid, 6-trifluoromethyl-nicotinic acid, 2-trifluoromethyl-5-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 6-trifluoromethyl, 2-trifluoromethyl pyridine-5-carboxylic acid, 6-trifluoromethyl-3-pyridinecarboxylic acid | |
| [O-]C(=O)C1=CC=C(N=C1)C(F)(F)F | |
| 190.10 | |
| 191.11 | |
| 6-(Trifluoromethyl)nicotinic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only