Learn More
6-(Trifluoromethyl)indole, 97%
CAS: 13544-43-9 | C9H6F3N | 185.149 g/mol
Supplier: Thermo Scientific Chemicals L17009MD
Description
6-(Trifluoromethyl)indole is used in pharmaceutical intermediates, liquid crystal intermediates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-(Trifluoromethyl)indole is used in pharmaceutical intermediates, liquid crystal intermediates
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 6-(Trifluoromethyl)indole | |
| 101°C to 105°C | |
| MFCD00272316 | |
| 1242785 | |
| Insoluble in water. | |
| C1=CC(=CC2=C1C=CN2)C(F)(F)F | |
| 185.149 | |
| 185.15 |
| 13544-43-9 | |
| C9H6F3N | |
| 250 mg | |
| 6-trifluoromethyl indole, 6-trifluoromethylindole, 6-trifluoromethyl-1h-indole, 1h-indole, 6-trifluoromethyl, pubchem1713, acmc-1bscm, intermediates-zcf02044, 6-trifluoromethyl-indole, ksc910g4n | |
| BPYBYPREOVLFED-UHFFFAOYSA-N | |
| 6-(trifluoromethyl)-1H-indole | |
| 2777523 | |
| 97% |
Chemical Identifiers
| 13544-43-9 | |
| 185.149 | |
| BPYBYPREOVLFED-UHFFFAOYSA-N | |
| 2777523 | |
| C1=CC(=CC2=C1C=CN2)C(F)(F)F |
| C9H6F3N | |
| MFCD00272316 | |
| 6-trifluoromethyl indole, 6-trifluoromethylindole, 6-trifluoromethyl-1h-indole, 1h-indole, 6-trifluoromethyl, pubchem1713, acmc-1bscm, intermediates-zcf02044, 6-trifluoromethyl-indole, ksc910g4n | |
| 6-(trifluoromethyl)-1H-indole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only