missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(Trifluoromethyl)indole, 97%
Description
6-(Trifluoromethyl)indole is used in pharmaceutical intermediates, liquid crystal intermediates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-(Trifluoromethyl)indole is used in pharmaceutical intermediates, liquid crystal intermediates
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
6-(Trifluoromethyl)indole is used in pharmaceutical intermediates, liquid crystal intermediates
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 101°C to 105°C |
| Quantity | 1 g |
| Beilstein | 1242785 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 185.15 |
| Percent Purity | 97% |
| Chemical Name or Material | 6-(Trifluoromethyl)indole |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only