missing translation for 'onlineSavingsMsg'
Learn More

6-(Trifluoromethyl)-3-pyridinecarboxaldehyde 98.0+%, TCI America™

Catalog No. T28385G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28385G 5 g
T28381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28385G Supplier TCI America Supplier No. T28385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 386704-12-7
Molecular Formula C7H4F3NO
Molecular Weight (g/mol) 175.11
MDL Number MFCD01862647
InChI Key MRPAGRCGPAXOGS-UHFFFAOYSA-N
Synonym 6-trifluoromethyl pyridine-3-carboxaldehyde, 6-trifluoromethyl nicotinaldehyde, 2-trifluoromethyl pyridine-5-carboxaldehyde, 6-trifluoromethyl pyridine-3-carbaldehyde, 5-formyl-2-trifluoromethyl pyridine, 3-pyridinecarboxaldehyde, 6-trifluoromethyl, 6-trifluoromethyl-pyridine-3-carbaldehyde, 6-trifluoromethyl-3-pyridinecarboxaldehyde, 2-trifluoromethyl pyridine-5-carbaldehyde, pubchem15866
PubChem CID 2777762
IUPAC Name 6-(trifluoromethyl)pyridine-3-carbaldehyde
SMILES FC(F)(F)C1=NC=C(C=O)C=C1

Specifications

Melting Point 58°C
Color White-Yellow
Quantity 5 g
Formula Weight 175.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-(Trifluoromethyl)-3-pyridinecarboxaldehyde
TSCA No
Recommended Storage Refrigerator