missing translation for 'onlineSavingsMsg'
Learn More

(6-Thien-2-ylpyridin-3-yl)methylamine dihydrochloride, 90%, Thermo Scientific™

Catalog No. CC54863CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC54863CB 250mg Amber Glass Bottle
CC54863DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC54863CB Supplier Thermo Scientific Chemicals Supplier No. CC54863CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 913830-32-7
Molecular Formula C10H12Cl2N2S
Molecular Weight (g/mol) 263.18
MDL Number MFCD09025878
InChI Key FWNBHEVGWIALSL-UHFFFAOYSA-N
Synonym 6-thien-2-ylpyridin-3-yl methylamine dihydrochloride, 6-thiophen-2-yl pyridin-3-yl methanamine dihydrochloride, 1-6-thiophen-2-yl pyridin-3-yl methanamine dihydrochloride, 6-thiophen-2-yl pyrid-3-ylmethylamine dihydrochloride, 5-aminomethyl-2-thien-2-yl pyridine dihydrochloride, 6-2-thienyl-3-pyridyl methylamine, chloride, chloride, 2-5-aminomethyl pyridin-2-yl thiophene dihydrochloride, 3-pyridinemethanamine,6-2-thienyl-,hydrochloride 1:2, 1-6-thiophen-2-yl pyridin-3-yl methanamine-hydrogen chloride 1/2
PubChem CID 24229545
IUPAC Name (6-thiophen-2-ylpyridin-3-yl)methanamine;dihydrochloride
SMILES C1=CSC(=C1)C2=NC=C(C=C2)CN.Cl.Cl

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 263.19
Percent Purity 90%
Chemical Name or Material (6-Thien-2-ylpyridin-3-yl)methylamine dihydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.