missing translation for 'onlineSavingsMsg'
Learn More

(6-Thien-2-ylpyrid-3-yl)methanol, 95%, Thermo Scientific™

Chemical Identifiers

CAS 198078-57-8
Molecular Formula C10H9NOS
Molecular Weight (g/mol) 191.248
MDL Number MFCD06410427
InChI Key OLHQRPWNQRBQMA-UHFFFAOYSA-N
Synonym 6-thien-2-ylpyrid-3-yl methanol, 6-thiophen-2-yl pyridin-3-yl methanol, 3-pyridinemethanol,6-2-thienyl, 2-thien-2-yl-5-hydroxymethylpyridine, 6-2-thienyl-3-pyridyl methan-1-ol
PubChem CID 15411145
IUPAC Name (6-thiophen-2-ylpyridin-3-yl)methanol
SMILES C1=CSC(=C1)C2=NC=C(C=C2)CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC54809CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

198078-57-8
191.248
OLHQRPWNQRBQMA-UHFFFAOYSA-N
15411145
C1=CSC(=C1)C2=NC=C(C=C2)CO
C10H9NOS
MFCD06410427
6-thien-2-ylpyrid-3-yl methanol, 6-thiophen-2-yl pyridin-3-yl methanol, 3-pyridinemethanol,6-2-thienyl, 2-thien-2-yl-5-hydroxymethylpyridine, 6-2-thienyl-3-pyridyl methan-1-ol
(6-thiophen-2-ylpyridin-3-yl)methanol

Specifications

198078-57-8
100.0
C10H9NOS
6-thien-2-ylpyrid-3-yl methanol, 6-thiophen-2-yl pyridin-3-yl methanol, 3-pyridinemethanol,6-2-thienyl, 2-thien-2-yl-5-hydroxymethylpyridine, 6-2-thienyl-3-pyridyl methan-1-ol
C1=CSC(=C1)C2=NC=C(C=C2)CO
191.248
191.25
(6-Thien-2-ylpyrid-3-yl)methanol
94.5
95%
MFCD06410427
OLHQRPWNQRBQMA-UHFFFAOYSA-N
(6-thiophen-2-ylpyridin-3-yl)methanol
15411145
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents