missing translation for 'onlineSavingsMsg'
Learn More

6-Thien-2-ylnicotinaldehyde, 97%, Thermo Scientific™

Catalog No. p-3522562
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC54804CB 250mg
CC54804DA 1g
CC54804DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC54804CB Supplier Thermo Scientific Chemicals Supplier No. CC54804CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 834884-61-6
Molecular Formula C10H7NOS
Molecular Weight (g/mol) 189.232
MDL Number MFCD04115421
InChI Key TZMYWLDEGHJHDI-UHFFFAOYSA-N
Synonym 6-thien-2-ylnicotinaldehyde, 6-thiophen-2-yl nicotinaldehyde, 6-thiophen-2-yl pyridine-3-carbaldehyde, 6-thiophen-2-yl-pyridine-3-carbaldehyde, 6-thien-2-ylnicotinylaldehyde, 6-2-thienyl pyridine-3-carbaldehyde, 6-thien-2-ylpyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde,6-2-thienyl, 2-thien-2-ylpyridine-5-carboxaldehyde
PubChem CID 24229542
IUPAC Name 6-thiophen-2-ylpyridine-3-carbaldehyde
SMILES C1=CSC(=C1)C2=NC=C(C=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250mg
Formula Weight 189.24
Percent Purity 97%
Chemical Name or Material 6-Thien-2-ylnicotinaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.