missing translation for 'onlineSavingsMsg'
Learn More

6-Thien-2-ylnicotinaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC54804DE

Quantity 5g

 View more versions of this product

Catalog No. CC54804DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

834884-61-6
189.232
TZMYWLDEGHJHDI-UHFFFAOYSA-N
24229542
C1=CSC(=C1)C2=NC=C(C=C2)C=O
C10H7NOS
MFCD04115421
6-thien-2-ylnicotinaldehyde, 6-thiophen-2-yl nicotinaldehyde, 6-thiophen-2-yl pyridine-3-carbaldehyde, 6-thiophen-2-yl-pyridine-3-carbaldehyde, 6-thien-2-ylnicotinylaldehyde, 6-2-thienyl pyridine-3-carbaldehyde, 6-thien-2-ylpyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde,6-2-thienyl, 2-thien-2-ylpyridine-5-carboxaldehyde
6-thiophen-2-ylpyridine-3-carbaldehyde

Specifications

6-Thien-2-ylnicotinaldehyde
96.5
97%
5g
6-thien-2-ylnicotinaldehyde, 6-thiophen-2-yl nicotinaldehyde, 6-thiophen-2-yl pyridine-3-carbaldehyde, 6-thiophen-2-yl-pyridine-3-carbaldehyde, 6-thien-2-ylnicotinylaldehyde, 6-2-thienyl pyridine-3-carbaldehyde, 6-thien-2-ylpyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde,6-2-thienyl, 2-thien-2-ylpyridine-5-carboxaldehyde
C1=CSC(=C1)C2=NC=C(C=C2)C=O
189.232
189.24
834884-61-6
100.0
C10H7NOS
MFCD04115421
TZMYWLDEGHJHDI-UHFFFAOYSA-N
6-thiophen-2-ylpyridine-3-carbaldehyde
24229542
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents