missing translation for 'onlineSavingsMsg'
Learn More

6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester, 97%

Catalog No. p-4813240
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444660010 1 g
AC444660050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC444660010 Supplier Thermo Scientific Chemicals Supplier No. 444660010
Only null left

CAS: 910462-31-6 | C16H25BN2O4 | 320.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 910462-31-6
Molecular Formula C16H25BN2O4
Molecular Weight (g/mol) 320.20
MDL Number MFCD07781162
InChI Key USZVCDRKIVKICD-UHFFFAOYSA-N
Synonym tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 2-tert-butoxycarbonylamino pyridine-5-boronic acid, pinacol ester, 2-boc-amino pyridine-5-boronic acid, pinacol ester, 2-boc-amino pyridine-5-boronic acid pinacol ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ylcarbamate, boc-6-aminopyridine-3-boronic acid pinacol ester, carbamic acid,n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl-,1,1-dimethylethyl ester, tert-butyl n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate
PubChem CID 17750222
IUPAC Name tert-butyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 203.0°C to 205.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 320.2
Percent Purity 97%
Chemical Name or Material 6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only