missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Quinolinol 96.0+%, TCI America™
$130.48 - $516.35
Chemical Identifiers
| CAS | 580-16-5 |
|---|---|
| Molecular Formula | C9H7NO |
| Molecular Weight (g/mol) | 145.161 |
| MDL Number | MFCD00047611 |
| InChI Key | OVYWMEWYEJLIER-UHFFFAOYSA-N |
| Synonym | 6-hydroxyquinoline, 6-quinolinol, 1h-1,6-epoxyquinoline, ccris 4331, 6-chinolinol, 6-hydroxyquinoline 6-quinolinol, quinoline-6-ol, 6-quinolinoloxine, 6-hydroxy-chinolin, 6-hydroxy-quinoline |
| PubChem CID | 11374 |
| ChEBI | CHEBI:48994 |
| IUPAC Name | quinolin-6-ol |
| SMILES | C1=CC2=C(C=CC(=C2)O)N=C1 |
Chemical Identifiers
| 580-16-5 | |
| 145.161 | |
| OVYWMEWYEJLIER-UHFFFAOYSA-N | |
| 11374 | |
| quinolin-6-ol |
| C9H7NO | |
| MFCD00047611 | |
| 6-hydroxyquinoline, 6-quinolinol, 1h-1,6-epoxyquinoline, ccris 4331, 6-chinolinol, 6-hydroxyquinoline 6-quinolinol, quinoline-6-ol, 6-quinolinoloxine, 6-hydroxy-chinolin, 6-hydroxy-quinoline | |
| CHEBI:48994 | |
| C1=CC2=C(C=CC(=C2)O)N=C1 |
Specifications
| 580-16-5 | |
| MFCD00047611 | |
| 6-hydroxyquinoline, 6-quinolinol, 1h-1,6-epoxyquinoline, ccris 4331, 6-chinolinol, 6-hydroxyquinoline 6-quinolinol, quinoline-6-ol, 6-quinolinoloxine, 6-hydroxy-chinolin, 6-hydroxy-quinoline | |
| C1=CC2=C(C=CC(=C2)O)N=C1 | |
| 145.161 | |
| CHEBI:48994 | |
| ≥96.0% (T) | |
| 6-Quinolinol |
| C9H7NO | |
| 5 g | |
| OVYWMEWYEJLIER-UHFFFAOYSA-N | |
| quinolin-6-ol | |
| 11374 | |
| 145.16 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : VC4130000
TSCA : No