missing translation for 'onlineSavingsMsg'
Learn More

6-Quinolineboronic acid pinacol ester, 97%

Catalog No. AC439640050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC439640050 5 g
AC439640010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC439640050 Supplier Thermo Scientific Chemicals Supplier No. 439640050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 406463-06-7 | C15H18BNO2 | 255.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 406463-06-7
Molecular Formula C15H18BNO2
Molecular Weight (g/mol) 255.12
MDL Number MFCD06411327
InChI Key VMFALDWCEQUFSX-UHFFFAOYSA-N
Synonym 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 6-quinolineboronic acid pinacol ester, quinoline-6-boronic acid pinacol ester, 6-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline-6-boronic acid, pinacol ester, 6-quinolylboronic acid pinacol ester, quinoline-5-boronic acid pinacol ester, 2-quinolin-6-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane-2-yl quinoline
PubChem CID 16217784
IUPAC Name 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
SMILES CC1(C)OB(OC1(C)C)C1=CC2=CC=CN=C2C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 65°C to 67°C
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 255.13
Percent Purity 97%
Physical Form Crystalline Solid or Powder
Chemical Name or Material 6-Quinolineboronic acid pinacol ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only