Learn More
6-Pyrrolidin-1-ylpyridine-2-carboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 450368-20-4 |
|---|---|
| Molecular Formula | C10H12N2O2 |
| Molecular Weight (g/mol) | 192.22 |
| MDL Number | MFCD09064946 |
| InChI Key | HXPHLYQBFPDHNK-UHFFFAOYSA-N |
| Synonym | 6-1-pyrrolidyl pyridine-2-carboxylic acid, 6-pyrrolidin-1-yl picolinic acid, 6-pyrrolidin-1-yl pyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 6-1-pyrrolidinyl, 6-pyrrolidinylpyridine-2-carboxylic acid, 6-1-pyrrolidyl pyridine-2-carboxylicacid, 2-pyridinecarboxylicacid,6-1-pyrrolidinyl, 6-1-pyrrolidinyl-2-pyridinecarboxylic acid |
| PubChem CID | 23509740 |
| IUPAC Name | 6-(pyrrolidin-1-yl)pyridine-2-carboxylic acid |
| SMILES | OC(=O)C1=NC(=CC=C1)N1CCCC1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
CC38001CB
|
N/A | 250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC38001DA
|
N/A | 1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 450368-20-4 | |
| 192.22 | |
| HXPHLYQBFPDHNK-UHFFFAOYSA-N | |
| 23509740 | |
| OC(=O)C1=NC(=CC=C1)N1CCCC1 |
| C10H12N2O2 | |
| MFCD09064946 | |
| 6-1-pyrrolidyl pyridine-2-carboxylic acid, 6-pyrrolidin-1-yl picolinic acid, 6-pyrrolidin-1-yl pyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 6-1-pyrrolidinyl, 6-pyrrolidinylpyridine-2-carboxylic acid, 6-1-pyrrolidyl pyridine-2-carboxylicacid, 2-pyridinecarboxylicacid,6-1-pyrrolidinyl, 6-1-pyrrolidinyl-2-pyridinecarboxylic acid | |
| 6-(pyrrolidin-1-yl)pyridine-2-carboxylic acid |
Specifications
| 450368-20-4 | |
| 100.0 | |
| C10H12N2O2 | |
| 6-1-pyrrolidyl pyridine-2-carboxylic acid, 6-pyrrolidin-1-yl picolinic acid, 6-pyrrolidin-1-yl pyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 6-1-pyrrolidinyl, 6-pyrrolidinylpyridine-2-carboxylic acid, 6-1-pyrrolidyl pyridine-2-carboxylicacid, 2-pyridinecarboxylicacid,6-1-pyrrolidinyl, 6-1-pyrrolidinyl-2-pyridinecarboxylic acid | |
| OC(=O)C1=NC(=CC=C1)N1CCCC1 | |
| 192.22 | |
| 192.22 | |
| 6-Pyrrolidin-1-ylpyridine-2-carboxylic acid |
| 96.5 | |
| 97% | |
| MFCD09064946 | |
| HXPHLYQBFPDHNK-UHFFFAOYSA-N | |
| 6-(pyrrolidin-1-yl)pyridine-2-carboxylic acid | |
| 23509740 | |
| 97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning