missing translation for 'onlineSavingsMsg'
Learn More

6-Pyrrolidin-1-ylpyridine-2-carbonitrile, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC38016DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC38016DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

160017-13-0
173.22
KUVMTKFDWSRABZ-UHFFFAOYSA-N
10329785
N#CC1=NC(=CC=C1)N1CCCC1
C10H11N3
MFCD09064948
6-pyrrolidin-1-yl pyridine-2-carbonitrile, 2-pyridinecarbonitrile,6-1-pyrrolidinyl, 6-pyrrolidin-1-yl picolinonitrile, 6-pyrrolidinylpyridine-2-carbonitrile, 6-1-pyrrolidinyl-2-pyridinecarbonitrile
6-(pyrrolidin-1-yl)pyridine-2-carbonitrile

Specifications

6-Pyrrolidin-1-ylpyridine-2-carbonitrile
96.5
97%
C10H11N3
MFCD09064948
KUVMTKFDWSRABZ-UHFFFAOYSA-N
6-(pyrrolidin-1-yl)pyridine-2-carbonitrile
10329785
97%
160017-13-0
100.0
Amber Glass Bottle
5g
6-pyrrolidin-1-yl pyridine-2-carbonitrile, 2-pyridinecarbonitrile,6-1-pyrrolidinyl, 6-pyrrolidin-1-yl picolinonitrile, 6-pyrrolidinylpyridine-2-carbonitrile, 6-1-pyrrolidinyl-2-pyridinecarbonitrile
N#CC1=NC(=CC=C1)N1CCCC1
173.22
173.22
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents