missing translation for 'onlineSavingsMsg'
Learn More

6-Pyrrolidin-1-ylpyridine-2-carbonitrile, 97%, Thermo Scientific™

Catalog No. CC38016DA
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC38016DA 1g Amber Glass Bottle
CC38016DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC38016DA Supplier Thermo Scientific Chemicals Supplier No. CC38016DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 160017-13-0
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.22
MDL Number MFCD09064948
InChI Key KUVMTKFDWSRABZ-UHFFFAOYSA-N
Synonym 6-pyrrolidin-1-yl pyridine-2-carbonitrile, 2-pyridinecarbonitrile,6-1-pyrrolidinyl, 6-pyrrolidin-1-yl picolinonitrile, 6-pyrrolidinylpyridine-2-carbonitrile, 6-1-pyrrolidinyl-2-pyridinecarbonitrile
PubChem CID 10329785
IUPAC Name 6-pyrrolidin-1-ylpyridine-2-carbonitrile
SMILES N#CC1=NC(=CC=C1)N1CCCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 6-Pyrrolidin-1-ylpyridine-2-carbonitrile
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.