missing translation for 'onlineSavingsMsg'
Learn More

(6-Piperidinopyrid-2-yl)methylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 868755-51-5
Molecular Formula C11H17N3
Molecular Weight (g/mol) 191.28
MDL Number MFCD08690243
InChI Key AMSLKMQNMJEDRH-UHFFFAOYSA-N
Synonym 6-piperidinopyrid-2-yl methylamine, 6-piperidin-1-yl pyridin-2-yl methanamine, 1-6-piperidin-1-yl pyridin-2-yl methanamine, 6-piperidyl-2-pyridyl methylamine, 2-pyridinemethanamine,6-1-piperidinyl, 6-piperidin-1-yl pyridin-2-yl methylamine, 1-6-piperidin-1-yl pyridin-2-yl methylamine
PubChem CID 18525734
IUPAC Name (6-piperidin-1-ylpyridin-2-yl)methanamine
SMILES NCC1=NC(=CC=C1)N1CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC38113CB
View Documents
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC38113DA
View Documents
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

868755-51-5
191.28
AMSLKMQNMJEDRH-UHFFFAOYSA-N
18525734
NCC1=NC(=CC=C1)N1CCCCC1
C11H17N3
MFCD08690243
6-piperidinopyrid-2-yl methylamine, 6-piperidin-1-yl pyridin-2-yl methanamine, 1-6-piperidin-1-yl pyridin-2-yl methanamine, 6-piperidyl-2-pyridyl methylamine, 2-pyridinemethanamine,6-1-piperidinyl, 6-piperidin-1-yl pyridin-2-yl methylamine, 1-6-piperidin-1-yl pyridin-2-yl methylamine
(6-piperidin-1-ylpyridin-2-yl)methanamine

Specifications

868755-51-5
100.0
Amber Glass Bottle
MFCD08690243
AMSLKMQNMJEDRH-UHFFFAOYSA-N
(6-piperidin-1-ylpyridin-2-yl)methanamine
18525734
97%
96.5
97%
C11H17N3
6-piperidinopyrid-2-yl methylamine, 6-piperidin-1-yl pyridin-2-yl methanamine, 1-6-piperidin-1-yl pyridin-2-yl methanamine, 6-piperidyl-2-pyridyl methylamine, 2-pyridinemethanamine,6-1-piperidinyl, 6-piperidin-1-yl pyridin-2-yl methylamine, 1-6-piperidin-1-yl pyridin-2-yl methylamine
NCC1=NC(=CC=C1)N1CCCCC1
191.28
191.28
(6-Piperidinopyrid-2-yl)methylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents