missing translation for 'onlineSavingsMsg'
Learn More

6-Phenylhexanoic Acid 98.0+%, TCI America™

Catalog No. P11085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11085G 5 g
P110825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11085G Supplier TCI America Supplier No. P11085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5581-75-9
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00014381
InChI Key JTXZPQIXIXYMDY-UHFFFAOYSA-N
Synonym benzenehexanoic acid, unii-43u2r7t4ec, 6-phenylcaproic acid, phenyl hexanoic acid, 6-phenylhexanoicacid, 6-phenyl-hexanoic acid, acmc-1ap1s, 6-phenylhexanoic acid
PubChem CID 79695
IUPAC Name 6-phenylhexanoic acid
SMILES C1=CC=C(C=C1)CCCCCC(=O)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 192.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 6-Phenylhexanoic Acid
TSCA No
Recommended Storage Refrigerator