missing translation for 'onlineSavingsMsg'
Learn More

6-Phenylhexanoic Acid 98.0+%, TCI America™

Supplier:  TCI America P110825G

Quantity 25 g

 View more versions of this product

Catalog No. P110825G


Only null left

Description

Description

Specifications

Chemical Identifiers

5581-75-9
192.258
JTXZPQIXIXYMDY-UHFFFAOYSA-N
79695
C1=CC=C(C=C1)CCCCCC(=O)O
C12H16O2
MFCD00014381
benzenehexanoic acid, unii-43u2r7t4ec, 6-phenylcaproic acid, phenyl hexanoic acid, 6-phenylhexanoicacid, 6-phenyl-hexanoic acid, acmc-1ap1s, 6-phenylhexanoic acid
6-phenylhexanoic acid

Specifications

6-Phenylhexanoic Acid
Yellow
25 g
benzenehexanoic acid, unii-43u2r7t4ec, 6-phenylcaproic acid, phenyl hexanoic acid, 6-phenylhexanoicacid, 6-phenyl-hexanoic acid, acmc-1ap1s, 6-phenylhexanoic acid
C1=CC=C(C=C1)CCCCCC(=O)O
192.258
192.26
Liquid
5581-75-9
C12H16O2
MFCD00014381
JTXZPQIXIXYMDY-UHFFFAOYSA-N
6-phenylhexanoic acid
79695
≥98.0% (GC,T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions