missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Phenyl-3(2H)-pyridazinone 98.0+%, TCI America™
$238.60 - $724.75
Chemical Identifiers
| CAS | 2166-31-6 |
|---|---|
| Molecular Formula | C10H8N2O |
| Molecular Weight (g/mol) | 172.187 |
| MDL Number | MFCD00114953 |
| InChI Key | IJUIPRDMWWBTTQ-UHFFFAOYSA-N |
| Synonym | 3-Hydroxy-6-phenylpyridazine |
| PubChem CID | 75106 |
| IUPAC Name | 3-phenyl-1H-pyridazin-6-one |
| SMILES | C1=CC=C(C=C1)C2=NNC(=O)C=C2 |
Chemical Identifiers
| 2166-31-6 | |
| 172.187 | |
| IJUIPRDMWWBTTQ-UHFFFAOYSA-N | |
| 75106 | |
| C1=CC=C(C=C1)C2=NNC(=O)C=C2 |
| C10H8N2O | |
| MFCD00114953 | |
| 3-Hydroxy-6-phenylpyridazine | |
| 3-phenyl-1H-pyridazin-6-one |
Specifications
| 2166-31-6 | |
| Yellow | |
| MFCD00114953 | |
| 3-Hydroxy-6-phenylpyridazine | |
| C1=CC=C(C=C1)C2=NNC(=O)C=C2 | |
| 172.187 | |
| 172.19 | |
| Crystalline Powder |
| 203°C | |
| C10H8N2O | |
| 1 g | |
| IJUIPRDMWWBTTQ-UHFFFAOYSA-N | |
| 3-phenyl-1H-pyridazin-6-one | |
| 75106 | |
| ≥98.0% (GC) | |
| 6-Phenyl-3(2H)-pyridazinone |
Safety and Handling
TSCA : No