missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Phenyl-2-thiouracil 98.0+%, TCI America™
$223.68 - $671.06
Chemical Identifiers
| CAS | 36822-11-4 |
|---|---|
| Molecular Formula | C10H8N2OS |
| Molecular Weight (g/mol) | 204.247 |
| MDL Number | MFCD00059166 |
| InChI Key | XEKNACRTWJHOCE-UHFFFAOYSA-N |
| PubChem CID | 697057 |
| IUPAC Name | 6-phenyl-2-sulfanylidene-1H-pyrimidin-4-one |
| SMILES | C1=CC=C(C=C1)C2=CC(=O)NC(=S)N2 |
Chemical Identifiers
| 36822-11-4 | |
| 204.247 | |
| XEKNACRTWJHOCE-UHFFFAOYSA-N | |
| 6-phenyl-2-sulfanylidene-1H-pyrimidin-4-one |
| C10H8N2OS | |
| MFCD00059166 | |
| 697057 | |
| C1=CC=C(C=C1)C2=CC(=O)NC(=S)N2 |
Specifications
| 36822-11-4 | |
| C10H8N2OS | |
| 1 g | |
| C1=CC=C(C=C1)C2=CC(=O)NC(=S)N2 | |
| 204.247 | |
| 204.25 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00059166 | |
| XEKNACRTWJHOCE-UHFFFAOYSA-N | |
| 6-phenyl-2-sulfanylidene-1H-pyrimidin-4-one | |
| 697057 | |
| ≥98.0% (HPLC,T) | |
| 6-Phenyl-2-thiouracil |
Safety and Handling
TSCA : No